Arteriosclerosis, thrombosis, and vascular biology
Statins upregulate PCSK9, the gene encoding the proprotein convertase neural apoptosis-regulated convertase-1 implicated in familial hypercholesterolemia.
PMID 15178557
Neural apoptosis-regulated convertase (NARC)-1 is the newest member of the proprotein convertase family implicated in the cleavage of a variety of protein precursors. The NARC-1 gene, PCSK9, has been identified recently as the third locus implicated in autosomal dominant hypercholesterolemia (ADH). The 2 other known genes implicated in ADH encode the low-density lipoprotein receptor and apolipoprotein B. As an approach toward the elucidation of the physiological role(s) of NARC-1, we studied its transcriptional regulation. Using quantitative RT-PCR, we assessed NARC-1 regulation under conditions known to regulate genes involved in cholesterol metabolism in HepG2 cells and in human primary hepatocytes. We found that NARC-1 expression was strongly induced by statins in a dose-dependent manner and that this induction was efficiently reversed by mevalonate. NARC-1 mRNA level was increased by cholesterol depletion but insensitive to liver X receptor activation. Human, mouse, and rat PCSK9 promoters contain 2 typical conserved motifs for cholesterol regulation: a sterol regulatory element (SRE) and an Sp1 site. PCSK9 regulation is typical of that of the genes implicated in lipoprotein metabolism. In vivo, PCSK9 is probably a target of SRE-binding protein (SREBP)-2.
Related Materials