EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Crystal structure communications
The supramolecular structure of 6-hydroxy-1,3-benzoxathiol-2-one (tioxolone).
PMID 15178861
Abstract
The planar molecules of 6-hydroxy-1,3-benzoxathiol-2-one, C(7)H(4)O(3)S, are linked by extensive O-H.O and C-H.O hydrogen bonding and are further stablilized by face-to-face pi-pi interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217077
6-Hydroxy-1,3-benzoxathiol-2-one, ≥98%
C7H4O3S