EMAIL THIS PAGE TO A FRIEND
Cutis
Overview of cefdinir: pharmacokinetics, safety, and efficacy in the treatment of uncomplicated skin and skin structure infections.
PMID 15182160
Abstract
Uncomplicated skin and skin structure infections commonly are caused by Staphylococcus aureus and Streptococcus pyogenes. Cefdinir, an extended spectrum, third-generation cephalosporin is a safe and effective means of treating skin infections caused by these organisms, as well as many gram-negative pathogens. This article summarizes the pharmacokinetics, dosing schedule, adverse event profile, and efficacy data for cefdinir in adult and pediatric, populations in the treatment of uncomplicated skin and skin structure infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7118 Cefdinir, ≥95% (HPLC)
C14H13N5O5S2