EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Conversion of alkyltrifluoroborates into alkyldichloroboranes with tetrachlorosilane in coordinating solvents.
PMID 15188479