EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Purification and characterization of an acid phosphatase from the commercial mushroom Agaricus bisporus.
PMID 15188886
Abstract
Acid phosphatase [AP; EC 3.1.3.2], a key enzyme involved in the synthesis of mannitol in Agaricus bisporus, was purified to homogeneity and characterized. The native enzyme appeared to be a high molecular weight type glycoprotein. It has a molecular weight of 145 kDa and consists of four identical 39-kDa subunits. The isoelectric point of the enzyme was found at 4.7. Maximum activity occurred at 65 degrees C. The optimum pH range was between 3.5 and 5.5, with maximum activity at pH 4.75. The enzyme was unaffected by EDTA, and inhibited by tartrate and inorganic phosphate. The enzyme exhibits a Km for p-nitrophenylphosphate and fructose-6-phosphate of 370 microM and 3.1 mM, respectively. A broad substrate specificity was observed with significant activities for fructose-6-phosphate, glucose-6-phosphate, mannitol-1-phosphate, AMP and beta-glycerol phosphate. Only phosphomonoesters were dephosphorylated. Antibodies raised against the purified enzyme could precipitate AP activity from a cell-free extract in an anticatalytic immunoprecipitation test.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92416
D-Mannitol 1-phosphate lithium salt, ≥95% (TLC)
C6H15O9P · xLi+