EMAIL THIS PAGE TO A FRIEND
Food additives and contaminants
Development of an analytical method for 3-monochloropropane-1,2-diol in soy sauce using 4-heptanone as derivatizing agent.
PMID 15195467
Abstract
3-Monochloro-1,2-propane diol is a suspected carcinogen found in hydrolysed vegetable protein products such as soy sauce. A method is described for the analysis of 3-monochloro-1,2-propane diol in soy sauce by gas chromatography-mass spectrometry at a concentration range of 1-5000 ng g-1 using 4-heptanone as the derivatizing ketone and 3-monochloro-1,2-propane diol-d5 as the internal standard. The limit of detection for the method in the soy sauce matrix was 0.48 ng g-1 and the limit of quantification was 1.2 ng g-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W254606
4-Heptanone, ≥97%, FG
C7H14O
101745
4-Heptanone, 98%
C7H14O