EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
New antifungal activity of penicillic acid against Phytophthora species.
PMID 15195966
Abstract
Penicillic acid was isolated from a culture filtrate of Aspergillus sclerotiorum. It had a high in vitro antifungal activity against Phytophthora spp., which has not been previously reported. MICs of penicillic acid were from 1 to 25 microg ml(-1) against Phytophthora spp. Penicillic acid induced abnormal branch formation, apical branching, and swelling in P. capsici, in P. cactorum mycelia contained irregular branching and small spherical swelling at apices, in P. cambivora there was irregular branching and swelling, and in P. drechsleri there was irregular multiple spherical swelling at or near hyphal apices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0063
Penicillic acid, ≥98% (HPLC)
C8H10O4