EMAIL THIS PAGE TO A FRIEND
Chinese medical journal
Schistosoma japonicum: establishment of a mouse model that demonstrates concomitant immunity.
PMID 15198906
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108804
N-Allylthiourea, 98%
C4H8N2S
06064
N-Allylthiourea, purum, ≥98.0% (HPLC)
C4H8N2S