EMAIL THIS PAGE TO A FRIEND
Organic letters
Small peptides catalyze highly enantioselective direct aldol reactions of aldehydes with hydroxyacetone: unprecedented regiocontrol in aqueous media.
PMID 15200341
Abstract
[reaction: see text] L-Proline-based small peptides have been developed as efficient catalysts for the asymmetric direct aldol reactions of hydroxyacetone with aldehydes. Chiral 1,4-diols 7, which are disfavored products in similar aldol reactions catalyzed by either aldolases or L-proline, were obtained in high yields and enantioselectivities of up to 96% ee with peptides 3 and 4 in aqueous media.
Related Materials