EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Separation of capsular polysaccharide-K4- and defructosylated-K4-derived disaccharides by high-performance liquid chromatography and postcolumn derivatization with 2-cyanoacetamide and fluorimetric detection.
PMID 15203345
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108448
Cyanoacetamide, 99%
C3H4N2O