EMAIL THIS PAGE TO A FRIEND
Journal of obstetrics and gynaecology : the journal of the Institute of Obstetrics and Gynaecology
A prospective study of pain following loop excision of the transformation zone under local anaesthesia.
PMID 15203634
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000415 Felypressin, European Pharmacopoeia (EP) Reference Standard
C46H65N13O11S2