EMAIL THIS PAGE TO A FRIEND
Food additives and contaminants
Quinoxaline-2-carboxylic acid in pigs: criteria to distinguish between the illegal use of carbadox and environmental contamination.
PMID 15204531
Abstract
Carbadox cannot be used in food-producing animals within the European Union following the adoption of Commission Regulation EC 2788/98/EC. Monitoring of the longest remaining residue--quinoxaline-2-car-boxylic acid (QCA)--is the most effective way of enforcing the prohibition on its use. The study was under taken to determine if QCA could be passed from pig to pig following the exposure of unmedicated animals to housing that had previously contained medicated animals. Drug-withdrawal studies were also carried out on medicated animals. Distinction between treated animals and those exposed to QCA might be required by competent national authorities to determine whether a positive result for QCA in tissue is truly 'violative'. Comparison of the ratio concentrations of QCA in tissues and body fluids was made to determine if they, could be used as criteria for discrimination between illegally treated animals and environmental contamination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293407
2-Quinoxalinecarboxylic acid, 97%
C9H6N2O2