EMAIL THIS PAGE TO A FRIEND
Metal ions in biological systems
The effect of cytoprotective agents in platinum anticancer therapy.
PMID 15206103
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000317 Mesna impurity D, European Pharmacopoeia (EP) Reference Standard
C4H10O6S4