EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Highly efficient biocatalytic resolution of cis- and trans-3-aminoindan-1-ol: syntheses of enantiopure orthogonally protected cis- and trans-indane-1,3-diamine.
PMID 15214084
Abstract
The efficient chemoenzymatic synthesis of enantiopure 1,3-difunctionalized indane derivatives has been achieved. The corresponding cis and trans N-protected amino alcohols were successfully resolved by acetylation using lipase B, which is a biocatalyst isolated from Candida antarctica. All the possible isomers were obtained in very good chemical yields and ee values (>99 %). The utility of these compounds was subsequently shown by the preparation of orthogonally protected cis- and trans-indane-1,3-diamine using a Mitsunobu reaction. Both enantiomers of the trans isomer and a desymmetrized cis diastereomer were prepared in enantiopure form. Complete inversion of configuration during the Mitsunobu reaction was demonstrated by a combination of NMR techniques and molecular modeling. The utility and versatility of the strategy was also demonstrated by the selective deprotection of each nitrogen atom under mild reaction conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45789
Indan, analytical standard
C9H10
I1804
Indan, 95%
C9H10