EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Widespread expression of the AMPA receptor GluR2 subunit at glutamatergic synapses in the rat spinal cord and phosphorylation of GluR1 in response to noxious stimulation revealed with an antigen-unmasking method.
PMID 15215299
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB397
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4