EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Spinophilin blocks arrestin actions in vitro and in vivo at G protein-coupled receptors.
PMID 15218143
Abstract
Arrestin regulates almost all G protein-coupled receptor (GPCR)-mediated signaling and trafficking. We report that the multidomain protein, spinophilin, antagonizes these multiple arrestin functions. Through blocking G protein receptor kinase 2 (GRK2) association with receptor-Gbetagamma complexes, spinophilin reduces arrestin-stabilized receptor phosphorylation, receptor endocytosis, and the acceleration of mitogen-activated protein kinase (MAPK) activity following endocytosis. Spinophilin knockout mice were more sensitive than wild-type mice to sedation elicited by stimulation of alpha2 adrenergic receptors, whereas arrestin 3 knockout mice were more resistant, indicating that the signal-promoting, rather than the signal-terminating, roles of arrestin are more important for certain response pathways. The reciprocal interactions of GPCRs with spinophilin and arrestin represent a regulatory mechanism for fine-tuning complex receptor-orchestrated cell signaling and responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4532
(−)-N6-(2-Phenylisopropyl)adenosine, solid
C19H23N5O4