EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Protein kinase C phosphorylates caldesmon77 and vimentin and enhances albumin permeability across cultured bovine pulmonary artery endothelial cell monolayers.
PMID 1522136
Abstract
Cytoskeletal protein (CSP) interactions are critical to the contractile response in muscle and non-muscle cells. Current concepts suggest that activation of the contractile apparatus occurs through selective phosphorylation by specific cellular kinase systems. Because the Ca(2+)-phospholipid-dependent protein kinase C (PKC) is involved in the regulation of a number of key endothelial cell responses, the hypothesis that PKC modulates endothelial cell contraction and monolayer permeability was tested. Phorbol myristate acetate (PMA), a direct PKC activator, and alpha-thrombin, a receptor-mediated agonist known to increase endothelial cell permeability, both induced rapid, dose-dependent activation and translocation of PKC in bovine pulmonary artery endothelial cells (BPAEC), as assessed by gamma-[32P]ATP phosphorylation of H1 histone in cellular fractions. This activation was temporally associated with evidence of agonist-mediated endothelial cell contraction as demonstrated by characteristic changes in cellular morphology. Agonist-induced activation of the contractile apparatus was associated with increases in BPAEC monolayer permeability to albumin (approximately 200% increase with 10(-6) MPMA, approximately 400% increase with 10(-8) M alpha-thrombin). To more closely examine the role of PKC in activation of the contractile apparatus, PKC-mediated phosphorylation of two specific CSPs, the actin- and calmodulin-binding protein, caldesmon77, and the intermediate filament protein, vimentin, was assessed. In vitro phosphorylation of both caldesmon and vimentin was demonstrated by addition of exogenous, purified BPAEC PKC to unstimulated BPAEC homogenates, to purified bovine platelet caldesmon77, or to purified smooth muscle caldesmon150. Caldesmon77 and vimentin phosphorylation were observed in intact [32P]-labeled BPAEC monolayers stimulated with either PMA or alpha-thrombin, as detected by immunoprecipitation. In addition, BPAEC pretreatment with the PKC inhibitor, staurosporine, prevented alpha-thrombin- and PMA-induced phosphorylation of both cytoskeletal proteins, attenuated morphologic evidence of contraction, and abolished agonist-induced barrier dysfunction. These results demonstrate that agonist-stimulated PKC activity results in cytoskeletal protein phosphorylation in BPAEC monolayer, an event which occurs in concert with agonist-mediated endothelial cell contraction and resultant barrier dysfunction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
BPAEC, 86123101