EMAIL THIS PAGE TO A FRIEND
FEBS letters
VIGR--a novel inducible adhesion family G-protein coupled receptor in endothelial cells.
PMID 15225624
Abstract
Using a signal sequence trap for selection of differentially expressed secretory and membrane proteins, we identified a novel member of the adhesion family of G-protein coupled receptors (GPCRs), termed vascular inducible GPCR (VIGR). VIGR contains C1r-C1s, Uegf and Bmp1 (CUB) and pentraxin (PTX)-like modules and a mucin-like spacer, followed by seven transmembrane domains. By surface biotinylation as well as by immunofluorescence analysis we demonstrate that endogenous, highly glycosylated VIGR is expressed on the cell surface of endothelial cells (ECs) upon LPS or thrombin treatment, and inducible expression is mediated by MAP kinases, but not NF-kappaB. We show that VIGR is selectively expressed in ECs derived from larger vessels, but not from microvessels. In summary, VIGR represents a novel GPCR of the adhesion family, which is unique in its long extra-cellular domain comprising CUB and PTX-like modules and in its inducibility by LPS and thrombin in a subset of ECs, suggesting an important function in cell-adhesion and potentially links inflammation and coagulation.