EMAIL THIS PAGE TO A FRIEND
Nihon Kokyuki Gakkai zasshi = the journal of the Japanese Respiratory Society
[The present state of short course chemotherapy including pyrazinamide in Japan].
PMID 15228133
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4050000
Pyrazinamide, European Pharmacopoeia (EP) Reference Standard
C5H5N3O