EMAIL THIS PAGE TO A FRIEND
Archives of pathology & laboratory medicine
Phenytoin overview--metabolite interference in some immunoassays could be clinically important.
PMID 15229987
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161543
5-(4-Hydroxyphenyl)-5-phenylhydantoin, 98%
C15H12N2O3