EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Xanthohumol and related prenylflavonoids from hops and beer: to your good health!
PMID 15231405
Abstract
Xanthohumol (3'-[3,3-dimethyl allyl]-2',4',4-trihydroxy-6'-methoxychalcone) is the principal prenylated flavonoid of the female inflorescences of the hop plant ('hops'), an ingredient of beer. Human exposure to xanthohumol and related prenylflavonoids, such as 8-prenylnaringenin and isoxanthohumol, is primarily through beer consumption. Xanthohumol has been characterized a 'broad-spectrum' cancer chemopreventive agent in in vitro studies, while 8-prenylnaringenin enjoys fame as the most potent phytoestrogen known to date. These biological activities suggest that prenylflavonoids from hops have potential for application in cancer prevention programs and in prevention or treatment of (post-)menopausal 'hot flashes' and osteoporosis. Xanthohumol and 8-prenylnaringenin are metabolized into many flavonoid derivatives with modified 3,3-dimethyl allyl (prenyl) moieties. Xanthohumol is formed in lupulin glands by a specialized branch of flavonoid biosynthesis that involves prenylation and O-methylation of the polyketide intermediate chalconaringenin. Although a lupulin gland-specific chalcone synthase is known, the aromatic prenyltransferase and O-methyltransferase participating in xanthohumol have not been identified. The prenylflavonoid pathway is a possible target for breeding or biotechnological modification of hops with the aim of increasing xanthohumol levels for beer brewing and 8-prenylnaringenin levels for pharmaceutical production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01130595
Xanthohumol, primary reference standard
C21H22O5