EMAIL THIS PAGE TO A FRIEND
The Southeast Asian journal of tropical medicine and public health
Laboratory evaluation of five pesticides for control of Leptotrombidium fletcheri (Acari: Trombiculidae).
PMID 1523464
Abstract
Five pesticides were evaluated against laboratory colonies of Leptotrombidium fletcheri (Womersly and Heaslip) by the Pasteur pipet technique. The pesticides were dieldrin (LC50 = 3.6 ppm, LC99 = 18.2 ppm), bromopropylate (LC50 = 9.2 ppm, LC99 = 239.6 ppm), dicofol (LC50 = 27.8 ppm, LC99 = 118.1 ppm), fenthion (LC50 = 15.4 ppm, LC99 = 29.7 ppm), and malathion (LC50 = 84.7 ppm, LC99 = 313.9 ppm). Dieldrin was the most toxic. Dicofol was recommended for further evaluation in field trials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45357
Bromopropylate, PESTANAL®, analytical standard
C17H16Br2O3