EMAIL THIS PAGE TO A FRIEND
Molecular Biology of the Cell
Promoter-dependent roles for the Srb10 cyclin-dependent kinase and the Hda1 deacetylase in Tup1-mediated repression in Saccharomyces cerevisiae.
PMID 15240822