Description
Molecular Formula
Add to Cart
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A2153
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
A7906
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
A7030
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
A4503
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
A7888
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
A9647
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
A3294
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
A6003
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
A3059
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
A9418
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
A0281
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
A7638
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A8022
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3912
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3803
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
B4287
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
A8806
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
A4378
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
A7511
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
A4919
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
A2058
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
A3311
Bovine Serum Albumin, powder, BioXtra
A1933
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
A4161
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
A9430
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A2934
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
B6917
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
A3156
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
A9306
Bovine Serum Albumin, lyophilized powder, low endotoxin, ≥95% (agarose gel electrophoresis)