EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Abnormalities of presynaptic protein CDCrel-1 in striatum of rats reared in social isolation: relevance to neural connectivity in schizophrenia
PMID 15245502