EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Transverse dephasing optimised NMR spectroscopy in solids: natural-abundance 13C correlation spectra.
PMID 15253313
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151114
Cholesteryl acetate, 97%
C29H48O2
T4879
L-Tyrosine ethyl ester hydrochloride
C11H15NO3 · HCl