EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of novel spiro[2.3]hexane carbocyclic nucleosides via enzymatic resolution.
PMID 15255683
Abstract
[reaction: see text] Novel R- and S-spiro[2.3]hexane nucleosides have been synthesized. The key step involved the Pseudomonas cepacia lipase catalyzed resolution of racemic compound 2, synthesized in seven steps starting from diethoxyketene and diethyl fumarate, to give (+)-acetate 3 and (-)-alcohol 13. (+)-Acetate 3 and (-)-acetate 14 were converted to R- and S-9-(6-hydroxymethylspiro[2.3]hexane)-4-adenine, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47930
Diethyl fumarate, purum, ≥98.0% (GC)
C8H12O4
D95654
Diethyl fumarate, 98%
C8H12O4