EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Ferrocene-containing polyelectrolyte multilayer film-covered electrodes: electrocatalytic determination of ascorbic acid and use of inner blocking layers to improve the upper detection limit of the electrodes.
PMID 15257429
Abstract
A multilayer film composed of ferrocene(Fc)-appended poly(allylamine hydrochloride) (Fc-PAH) and poly(potassium vinylsulfate) (PVS) has been prepared on the surface of a gold(Au) electrode by using a layer-by-layer self-assembly technique. Fc-containing polyelectrolyte multilayer (PEM) film-modified electrodes can electrochemically catalyze the oxidation of ascorbic acid successfully. For a 2 (Fc-PAH/PVS) bilayer-covered electrode the catalytic current increased linearly with increasing concentration of ascorbic acid over the concentration range 6 micromol L(-1)-3 mmol L(-1). To extend the dynamic range for ascorbic acid, the surface of the Au electrode was first covered with a (PAH/PVS)(2) film on which an additional (Fc-PAH/PVS)(5) film was coated. This strategy successfully extended the dynamic range of the electrode up to 25 mmol L(-1) ascorbic acid, because the (PAH/PVS)(2) layer blocked access of ascorbic acid to the electrode surface. The upper detection limit of the (PAH/PVS)(2) (Fc-PAH/PVS)(5) film-modified electrode is much higher than those of Fc-based ascorbic acid sensors reported so far. Electron transfer is diffusion-controlled within the (PAH/PVS)(2)(Fc-PAH/PVS)(5) film.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
271969
Poly(vinyl sulfate) potassium salt, average Mw ~170,000