EMAIL THIS PAGE TO A FRIEND
Annales de dermatologie et de venereologie
[Fixed drug eruption induced by sulfaguanidine].
PMID 15258515
Abstract
Fixed drug eruption is a lesion induced by drugs. The family of drugs usually incriminated are sulfonamides, tetracyclines and pyrazols. We describe nine cases of fixed drug eruption induced by sulfaguanidine, a sulfonamide with local action. All the patients presented one or more fixed drug eruption reactivation lesions induced by sulfaguanidine as self-medication for diarrhea. The number of lesions increased in 7 cases after reactivation. The delay in occurrence of the fixed drug eruption decreased during the different episodes. The lesions predominated on the hands in 8 cases of 9. The sulfaguanidine must be added to the list of drug-induced fixed drug eruptions with limited absorption from the gastrointestinal tract.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32402
Sulfaguanidine, VETRANAL®, analytical standard
C7H10N4O2S
S8751
Sulfaguanidine
C7H10N4O2S