EMAIL THIS PAGE TO A FRIEND
Chemosphere
Chemical and enzymatic interactions of Direct Black 38 and Direct Brown 1 on release of carcinogenic amines.
PMID 15261529
Abstract
Release of amine products from azo compounds is of considerable interest, since most of the metabolized amine products have toxic and carcinogenic characters. Moreover, most of the azo dyes are extensively used as coloring agents in inks, textiles, leathers, food and pharmaceutical industries. The present study emphasis on the quantification and comparison of amines released from water soluble dyes by (i) extra cellular protein (ECP) of Streptomyces sp. SS07 and by (ii) chemical methods. It has been observed that both the methods release considerable quantities of similar type of amine products. Release of amine compounds by ECP and chemical reduction in acid and alkaline sweat medium from a leather garment sample was also assessed. ECP (0.7852 mg protein/mg of ECP) releases benzidine and 4-amino biphenyl from Direct Black 38 and Direct Brown 1 as stable products at pH 9.2 and at 37 degrees C for a contact period of 24 h. On comparison with chemical reduction, it was observed that about 5-20% increase in the release of amine products by ECP was observed. However, more than 60% of amine products were released by chemical method from leather garment samples than direct treatment with ECP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1144
Chlorazol Black
C34H25N9Na2O7S2