EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Molecular dynamics (MD) simulations for the prediction of chiral discrimination of N-acetylphenylalanine enantiomers by cyclomaltoheptaose (beta-cyclodextrin, beta-CD) based on the MM-PBSA (molecular mechanics-Poisson-Boltzmann surface area) approach.
PMID 15261589
Abstract
Molecular dynamics (MD) simulations were performed for the prediction of chiral discrimination of N-acetylphenylalanine enantiomers by cyclomaltoheptaose (beta-cyclodextrin, beta-CD). Binding free energies and various conformational properties were obtained using by the MM-PBSA (molecular mechanics Poisson-Boltzmann/surface area) approach. The calculated relative difference (DeltaDeltabinding) of binding free energy was in fine agreement with the experimentally determined value. The difference of rotameric distributions of guest N-acetylphenylalanine enantiomers complexed with the host, beta-CD, was observed after the conformational analyses, suggesting that the conformational changes of guest captured within host cavity would be a decisive factor for enantiodifferentiation at a molecular level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3876
N-Acetyl-D-phenylalanine
C11H13NO3
857459
N-Acetyl-L-phenylalanine, ReagentPlus®, 99%
C11H13NO3