EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Photolysis of octachloronaphthalene in hexane.
PMID 15266697
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442725 Octachloronaphthalene, analytical standard