EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Chemical constituents from Typhonium flagelliforme].
PMID 15272778
Abstract
To study the major chemical constituents from the root tuber of Typhyonium flagelliforme. Compounds were separated and purified with silica gel column and preparative HPLC, and their structures were elucidated on the basis of spectroscopic evidence (UV, IR, MS, NMR and 2D NMR). Four compounds were identified as 1-O-beta-glucopyranosyl-2-[(2-hydroxyloctadecanoyl) amido]-4,8-octadecadiene-1,3-diol (1), coniferin (2), beta-sitosterol (3) and beta-daucosterol (4). A cerebroside with significant antihepatotoxic activity and a phenylpropanoid glycoside were isolated from Typhyonium flagelliforme for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00103
Abietin, ≥90% (LC/MS-ELSD)
C16H22O8