EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica (Zagreb, Croatia)
Lipophilicity study of salicylamide.
PMID 15274753
Abstract
Molecular lipophilicity was studied using salicylamide as a model drug. Log P value for the target compound was experimentally determined by the shake-flask method and calculated using nine different computer programs based on atom/fragment contributions, structural parameters, atom-type electrotopological-state indices and neural network modeling, or topological structure descriptors. Our analysis demonstrates good agreement between the experimentally observed log P value of salicylamide and the value calculated by the CSLogP program, based on topological structure descriptors and electrotopological indices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
860417
Salicylamide, 99%
C7H7NO2
84230
Salicylamide, puriss., ≥99.0% (T)
C7H7NO2