EMAIL THIS PAGE TO A FRIEND
Current medicinal chemistry
Monoamine oxidases: certainties and uncertainties.
PMID 15279561
Abstract
A great deal has been learned about the behaviour of monoamine oxidase in the 75 years since it was first discovered, but there is still a great deal left to understand. This review concentrates on the dynamic aspects of our knowledge of the interactions of MAO with substrates and inhibitors and how it may collaborate with other enzymes, with particular emphasis on aspects that remain to be clarified.