EMAIL THIS PAGE TO A FRIEND
The Biochemical Journal
Sulpho-conjugation of ethanol in humans in vivo and by individual sulphotransferase forms in vitro.
PMID 15281911
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W255718
Hexanal, ≥97%, FCC, FG
C6H12O
W255726
Hexanal, natural, ≥95%, FG
C6H12O