EMAIL THIS PAGE TO A FRIEND
Prostate cancer and prostatic diseases
Long-term testosterone stimulation induces hyperplasia in the guinea-pig prostate.
PMID 15289812
Abstract
The relation between supraphysiologic circulating testosterone levels and prostatic diseases is unclear and difficult to study in men. Animal models may be advantageous. Based on a pilot study, testosterone enantate 50 mg (n=12) or 25 mg (n=12) was administered to guinea-pigs intramuscularly every 3 weeks, for either 7 or 14 months. The histopathology of the prostate was described. Epithelial hyperplasia was found in 14/21 animals receiving testosterone and in 7/12 very old animals, but no such changes were found in the sham or castrated animals. Testosterone stimulation seems to induce epithelial hyperplasia, but not cancer, in the guinea-pig prostate.