EMAIL THIS PAGE TO A FRIEND
Protein Expression and Purification
Decreases in yeast expression yields of the human adenosine A2a receptor are a result of translational or post-translational events.
PMID 15294291