EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
A novel amide N-glycoside of ansamitocins from Actinosynnema pretiosum.
PMID 15303496
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2836
Ansamitocin P-3 from Actinosynnema pretiosum, ≥90% (HPLC)
C32H43ClN2O9