EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Characterization of heparan sulphate 3-O-sulphotransferase isoform 6 and its role in assisting the entry of herpes simplex virus type 1.
PMID 15303968
Abstract
Heparan sulphate (HS) 3-O-sulphotransferase transfers sulphate to the 3-OH position of the glucosamine residue of HS to form 3-O-sulphated HS. The HS modified by 3-O-sulphotransferase isoform 3 binds to HSV-1 (herpes simplex virus type 1) gD (envelope glycoprotein D), and the resultant 3-O-sulphated HS serves as an entry receptor for HSV-1. In the present paper, we report the isolation and characterization of a novel HS 3-O-sulphotransferase isoform, designated HS 3-O-sulphotransferase isoform 6 (3-OST-6). Mouse 3-OST-6 gene was identified in the EST (expressed sequence tag) database and cloned into pcDNA3.1/Myc-His vector. A CHO (Chinese-hamster ovary) cell line that stably expresses 3-OST-6 (3OST6/CHO cells) was prepared. The disaccharide analysis of the HS isolated from 3OST6/CHO cells revealed that 3-OST-6 exhibits HS 3-O-sulphotransferase activity. Furthermore, 3OST6/CHO cells were susceptible to infection by HSV-1, but not by other alphaherpesviruses examined, suggesting that 3-OST-6 produces a specific entry receptor for HSV-1. Our results indicate that a new member of 3-OST family generates an entry receptor for HSV-1. The findings add to the growing body of evidence that HSV-1 entry is mediated by 3-O-sulphated HS generated by multiple members of 3-O-sulphotransferases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10312 CompoZr® Knockout ZFN Kit, Human HS3ST6 (NM_001009606)