EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Selective oxidation of alcohols at the benzylic position by benzeneseleninic anhydride.
PMID 15305004
Abstract
Benzeneseleninic anhydride in the presence of tert-butyl hydroperoxide in chlorobenzene at about 70 degrees C is an effective oxidizing agent for the selective oxidation of alcohols at the benzylic position.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
213012
Benzeneseleninic acid anhydride, 70%
C12H10O3Se2