EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Facile synthesis of oxabicyclic alkenes by ultrasonication-promoted diels-alder cycloaddition of furano dienes.
PMID 15307754
Abstract
Ultrasonic irradiation effectively promotes the Diels-Alder reaction of substituted furans with reactive dienophiles (i.e., dimethyl acetylenedicarboxylate (DMAD) and dimethyl maleate). Regiospecific furano Diels-Alder cycloaddition of 2-vinylic furans with DMAD furnished functionalized oxabicyclic alkenes in good yield under ultrasonication condition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238198
Dimethyl maleate, 96%
C6H8O4