EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A dinuclear, triple-stranded helicate with a diamide-bridged catechol/benzenedithiol ligand.
PMID 15317008
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270865
Benzene-1,2-dithiol, 96%
C6H6S2