EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
o-Aminoazotoluene.
PMID 15317117
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
121568
Fast Garnet GBC base, 97%
C14H15N3
31629
o-Aminoazotoluene, analytical standard
C14H15N3