EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
o-Anisidine hydrochloride.
PMID 15317122
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31597
o-Anisidine, analytical standard
C7H9NO
A88182
o-Anisidine, ≥99%
C7H9NO