EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Chlorendic acid.
PMID 15318386
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
411701
1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic acid, 99%
C9H4Cl6O4