EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Disperse Blue 1.
PMID 15323054
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215643
Disperse Blue 1, Dye content 30 %
C14H12N4O2