EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Lead acetate and lead phosphate.
PMID 15326666
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
663328
Lead hydrogen phosphate, 98%
HO4PPb