EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
4,4'-Methylenebis(N,N-dimethyl)benzenamine.
PMID 15326672
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M44451
4,4′-Methylenebis(N,N-dimethylaniline), 98%
C17H22N2