EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
o-Nitroanisole.
PMID 15326685
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
236284
2-Nitroanisole, ≥99%
C7H7NO3